Learn More
TARGETMOL CHEMICALS INC DMNQ 25MG
SDP
Catalog No. 502579768
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DMNQ is a 14-naphthoquinone that acts as a redox-cycling agent typically increasing intracellular superoxide and hydrogen peroxide formation.DMNQ increases ROS generationpurity: 99%
Catalog No. 50-257-9768 Supplier TARGETMOL CHEMICALS INC Supplier No. T542625MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DMNQ is a 14-naphthoquinone that acts as a redox-cycling agent typically increasing intracellular superoxide and hydrogen peroxide formation.DMNQ increases ROS generationpurity: 99%