Learn More
AdipoGen DMNQ
SDP
Supplier:  AdipoGen CDXD0205M010
Encompass_Preferred
Chemical. CAS 6956-96-3. Formula C12H10O4. MW 218.21. Synthetic A cell-permeable, non-alkylating, non-thiol, adduct-forming, redox cycling quinone. Generates superoxide production and induces cell proliferation, apoptosis and necrosis in vitro. Redox cycling agent.
Catalog No. 50-256-1186