Learn More
Abcam DMNQ, non-alkyiating redo x cycling quinone, 5MG
Catalog No. 502705546
Encompass_Preferred
MW 218.2 Da, Purity >99%. Cell-permeable, non-alkylating redox cycling quinone. Generates superoxide production and induces cell proliferation, apoptosis and necrosis in vitro. Redox cycling agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5546 Supplier Abcam Supplier No. AB1446265MG
MW 218.2 Da, Purity >99%. Cell-permeable, non-alkylating redox cycling quinone. Generates superoxide production and induces cell proliferation, apoptosis and necrosis in vitro. Redox cycling agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement