Learn More
Cayman Chemical Vicenin-2, Purity: ≥98%, CAS: 23666-13-9, 10 mg
SDP
Catalog No. 502946452
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; inhibits thrombin-induced aggregation of isolated mouse platelets from 2-30 µM; increases tail bleeding time in mice at 35.7 µg/animal; reduces tumor growth in a PC3 prostate cancer mouse xenograft model at 1 mg/kg; decreases trabecular separation, as well as increases trabecular thickness and area, in an ovariectomized rat model of osteoporosis; decreases ulceration and increases colon weight and length in a mouse model of DSS-induced colitis at 50 mg/kg
Catalog No. 50-294-6452 Supplier Cayman Chemical Supplier No. 3340310 MG
A flavonoid glycoside with diverse biological activities; inhibits thrombin-induced aggregation of isolated mouse platelets from 2-30 µM; increases tail bleeding time in mice at 35.7 µg/animal; reduces tumor growth in a PC3 prostate cancer mouse xenograft model at 1 mg/kg; decreases trabecular separation, as well as increases trabecular thickness and area, in an ovariectomized rat model of osteoporosis; decreases ulceration and increases colon weight and length in a mouse model of DSS-induced colitis at 50 mg/kg