Learn More
Cayman Chemical DMOG, 89464-63-1, 10 mg
SDP
Supplier:  Cayman Chemical 7121010
Encompass_Preferred
Molecular Formula C6H9NO5, Purity ≥98%, Formula Weight 175.1, Synonyms: N-(methoxyoxoacetyl)-glycine, methyl ester, Dimethyloxallyl Glycine; A cell permeable, competitive inhibitor of HIF-1 prolyl hydroxylase; stabilizes HIF-1 expression at normal oxygen tensions in cultured cells at concentrations between 0.1 and 1 mM.
Catalog No. NC0075934
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.