Learn More
Cayman Chemical DMOG, 89464-63-1, 100 mg
SDP
Supplier:  Cayman Chemical 71210100
Encompass_Preferred
A cell permeable, competitive inhibitor of HIF-1α prolyl hydroxylase; stabilizes HIF-1α expression at normal oxygen tensions in cultured cells at concentrations between 0.1 and 1 mM
Catalog No. NC1446696