Learn More
Medchemexpress LLC HY-15893 100mg , DMOG CAS:89464-63-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15893100MG
Encompass_Preferred
HY-15893 100mg DMOG CAS: 89464-63-1 DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal[4]. DMOG induces cell autophagy[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9761