Learn More
Apexbio Technology LLC DMOG 89464-63-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A45065.1
Encompass_Preferred
DMOG (Dimethyloxalylglycine CAS 89464-63-1) is a small molecule inhibitor of prolyl-4-hydroxylase domain (PHD) enzymes thereby modulating the stability of hypoxia-inducible factor (HIF) PHD inhibition by DMOG prevents HIF-1 prolyl hydroxylation and subsequent degradation stabilizing HIF-1 protein even under normoxic conditions (0 1 1 mmol/L in vitro) Additionally DMOG treatment suppresses NF- B signaling during LPS-induced inflammatory responses in animal models improving survival rates and enhancing IL-10 expression predominantly in peritoneal B-1 cells Thus DMOG serves as a valuable research tool to explore hypoxia signaling pathways and inflammatory responses
Catalog No. 50-000-61772