Learn More
TARGETMOL CHEMICALS INC DMOG 200MG
SDP
Catalog No. 502878595
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DMOG (Dimethyloxalylglycine), an antagonist of the alpha-ketoglutarate cofactor, is an inhibitor for HIF prolyl hydroxylase. Purity 98.26%
Catalog No. 50-287-8595 Supplier TARGETMOL CHEMICALS INC Supplier No. T1939200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DMOG (Dimethyloxalylglycine), an antagonist of the alpha-ketoglutarate cofactor, is an inhibitor for HIF prolyl hydroxylase. Purity 98.26%