Learn More
Medchemexpress LLC HY-15893 50mg Medchemexpress, DMOG CAS:89464-63-1 Purity:>98%
SDP
Catalog No. 502029179
Encompass_Preferred
Medchemexpress, HY-15893 50mg DMOG CAS:89464-63-1 DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal[2][4]. DMOG induces cell autophagy[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9179 Supplier Medchemexpress LLC Supplier No. HY1589350MG
Medchemexpress, HY-15893 50mg DMOG CAS:89464-63-1 DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal[2][4]. DMOG induces cell autophagy[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.