Learn More
Electron Microscopy Sciences DMP (2,2-Dimethoxypropane) 450 ML
SDP
Supplier:  Electron Microscopy Sciences 13281
Encompass_Preferred
(Acetone Dimethyl Acetal)
(CH₃)₂C(OCH₃)₂ F.W. 104.14
Used as dehydration agent for plant tissue in TEM. Lipid extracting properties of 2,2-Dimethoxypropane as revealed by electron microscopy and thin layer chromatography.
Catalog No. 50-980-363