Learn More
Electron Microscopy Sciences DMP-30 (2,4,6-Tri(dimethylaminomethyl) phenol) 25 ML
SDP
Supplier:  Electron Microscopy Sciences 13600
Encompass_Preferred
((2,4,6-Tri(dimethylaminomethyl) phenol))
C₁₅H₂₇N₃O
A tertiary amine, used as an accelerator for anhydride cures. One of the most popular accelerators available! Keep as dry as possible. Short shelf life. Use as fresh as possible. (2,4,6-Tri(dimethylaminomethyl) phenol)
Catalog No. 50-980-374