Learn More
Medchemexpress LLC HY-108590 5mg Medchemexpress, DMP-543 CAS:160588-45-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085905MG
Encompass_Preferred
Medchemexpress, HY-108590 5mg DMP-543 CAS:160588-45-4 DMP-543 (XR-543) is a KV7 channel blocker, also acts as a potent neurotransmitter release enhancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0748