Learn More
Medchemexpress LLC HY-12131 5mg Medchemexpress, DMP 696 CAS:202578-52-7 Purity:>98%
SDP
Catalog No. 502080965
Encompass_Preferred
Medchemexpress, HY-12131 5mg DMP 696 CAS:202578-52-7 DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist, used for the treatment of anxiety and depression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0965 Supplier Medchemexpress LLC Supplier No. HY121315MG
Medchemexpress, HY-12131 5mg DMP 696 CAS:202578-52-7 DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist, used for the treatment of anxiety and depression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.