Learn More
Echelon Biosciences Research Labs DMPC 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L1114250MG
Encompass
1,2-Dimyristoyl-sn-glycero-3-phosphocholine (DMPC) is a synthetic phospholipid used in liposomes and lipid bilayers for the study of biological membranes.Phosphatidylcholine (PC) is generally the most abundant lipid in animal cell membranes providing structural framework. PC is more common in the outer leaflet where it functions as part of the permeability barrier. PC is also the primary substrate of Phospholipase D enzymes which produce the signaling lipids phosphatidic acid and lysophosphatidic acid. This item is non returnable
Catalog No. 50-720-488