Learn More
Kyfora Bio DMPC 250 MG
Catalog No. 502559779
Encompass_Preferred
DMPC is a phosphatidylcholine (PC) and a zwitterionic synthetic phospholipid used to formulate lipid nanoparticles (LNPs), and small unilamellar vesicles (SUVs). PCs are the most abundant lipid in animal cell membranes and functions as part of the permeability barrier. DMPC has been used to study the properties of lipid monolayers and bilayers. Key application(s): Study of lipid bilayer, LNP synthesis, SUV synthesis.
Catalog No. 50-255-9779 Supplier Kyfora Bio Supplier No. 26447250
DMPC is a phosphatidylcholine (PC) and a zwitterionic synthetic phospholipid used to formulate lipid nanoparticles (LNPs), and small unilamellar vesicles (SUVs). PCs are the most abundant lipid in animal cell membranes and functions as part of the permeability barrier. DMPC has been used to study the properties of lipid monolayers and bilayers. Key application(s): Study of lipid bilayer, LNP synthesis, SUV synthesis.