Learn More
Echelon Biosciences Research Labs DMPE 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L2114250MG
Encompass
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine (DMPE) is a water soluble derivative of phosphatidylethanolamine with (14:0) myristic acid acyl chains. Phosphatidylethanolamine (PE) is formed primarily in the reaction of CDP-ethanolamine and diacylglycerol. PE is one of the major components of cell membranes in bacteria and in nervous system. It is useful in liposome production. This item is non returnable
Catalog No. 50-720-501