Learn More
Eurofins Discoverx DMPK, active, 250 µg. (14-649M)
Supplier:  Eurofins Discoverx 14649M
Encompass
6xHis-tagged recombinant, human DMPK amino acids 1-549. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9120