Learn More
Carnabio Usa Inc DMPK1(DMPK)/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0110510
Encompass
DMPK1(DMPK), Catalytic domain, Wild type, Amino Acid 1-544, AAA75236.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0752