Learn More
Dojindo Molecular Technologies Inc DMPO (1ML), CAS: 3317-61-1 is a spin trapping reagent for the study of free radicals. Some features include Ultra-high purity, greater S/N ratio, and no pre-purification required.
SDP
Supplier:  Dojindo Molecular Technologies Inc D04810
Encompass
Because of potential cancer risks and their age-promoting effects, free radicals in living bodies have become a frequently studied subject. DMPO is the most frequently used spin-trapping reagent for the study of free radicals. It is suitable for trapping oxygen radicals, especially superoxides, and for producing adducts with characteristic EPR (ESR) patterns. However, most commercially available DMPO contains impurities that cause high backgrounds. Thus, DMPO requires further purification for running experiments on EPR. The quality of Dojindo's DMPO is well controlled and Dojindo DMPO doesn’t require any pre-purification process. Thus, there are no impurities to cause a background problem.
Catalog No. NC0958234