Learn More
Enzo Life Sciences DMPO (1g). CAS: 3317-61-1
SDP
Supplier:  Enzo Life Sciences ALX430090G001
Encompass_Preferred
Cell permeable hydrophilic spin trap for both in vivo and in vitro studies of superoxide, O-, C-, S- and N- centered free radicals. Alternative name: 5,5-Dimethyl-1-pyrroline-N-oxide, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide. Purity: ≥98% (ELSD). Appearance: Pale yellow solid or oil. Solubility: Soluble in water, 100% ethanol, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9193