Learn More
TARGETMOL CHEMICALS INC DMPO 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T22739200MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DMPO (55-Dimethyl-1-pyrroline N-oxide) is a kind of water-soluble nitric oxide spin trap which allows the measurement of oxygen-centered free radicals in biological systems at room temperature using electron spin resonance. It has a high reaction rate constant for superoxide and hydroxyl radicals and distinguishes simultaneously among a variety of important biologically generated free radicals. purity: 99%
Catalog No. 50-258-0930