Learn More
Abcam DMPO, hydrophilic spin trap, 100MG
Catalog No. 502705569
Encompass_Preferred
MW 113.16 Da, Purity >99%. Cell-permeable hydrophilic spin trap. React with radicals forming stable adducts. Reacts with O-, N-, S-, and C-centred radicals. Reduces NMDA-induced neuronal death in vitro. Reduces nickel-induced DNA damage in vitro. Antioxidant and neuroprotective agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5569 Supplier Abcam Supplier No. AB144610100MG
MW 113.16 Da, Purity >99%. Cell-permeable hydrophilic spin trap. React with radicals forming stable adducts. Reacts with O-, N-, S-, and C-centred radicals. Reduces NMDA-induced neuronal death in vitro. Reduces nickel-induced DNA damage in vitro. Antioxidant and neuroprotective agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement