Learn More
Enzo Life Sciences DMPO monoclonal antibody (1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX803340C100
Encompass_Preferred
Clone: 1. Isotype: IgG. Immunogen: DMPO-octanoic acid conjugated to ovalbumin. Source: Hybridoma tissue culture supernatant. Species reactivity: Species independent. Specificity: Recognizes DMPO (ALX-430-090), DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Applications: ELISA, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 10% glycerol and 0.02% sodium azide.
Catalog No. 50-200-9740