Learn More
Medchemexpress LLC DMPQ dihydrochloride | 1123491-15-5 | 99.1% | 339.22 | 10 MG
SDP
Catalog No. 5000346281
Encompass_Preferred
DMPQ dihydrochloride is a potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ) with an IC50 of 80 nM. It is intended for research use only.
  • Potent and selective inhibitor of human PDGFRβ.
Catalog No. 50-003-46281 Supplier Medchemexpress LLC Supplier No. HY1086271MG
DMPQ dihydrochloride is a potent and selective inhibitor of human platelet-derived growth factor receptor β (PDGFRβ) with an IC50 of 80 nM. It is intended for research use only.
  • Potent and selective inhibitor of human PDGFRβ.