Learn More
Abcam DMPQ dihydrochloride, PDGFR beta inhibitor, 50MG
Catalog No. 502706735
Encompass_Preferred
MW 339.2 Da, Purity >99%. Potent and selective β-type platelet derived growth factor receptor tyrosine kinase (PDGFRβ) inhibitor (IC₅₀ = 80 nM). Shows >100-fold selectivity over EGFR, PKA, PKC, p56 and erbB2. Also available in simple stock solutions (ab146732) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6735 Supplier Abcam Supplier No. AB14126950MG
Add to Cart
Add to Cart
MW 339.2 Da, Purity >99%. Potent and selective β-type platelet derived growth factor receptor tyrosine kinase (PDGFRβ) inhibitor (IC₅₀ = 80 nM). Shows >100-fold selectivity over EGFR, PKA, PKC, p56 and erbB2. Also available in simple stock solutions (ab146732) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement