Learn More
Cayman Chemical DMPQ (hydrochloride), Purity: ≥98%, CAS: 1123491-15-5, 10 mg
SDP
Catalog No. 502965080
Encompass_Preferred
A potent inhibitor of human PDGFRβ (IC50 = 80 nM)
Catalog No. 50-296-5080 Supplier Cayman Chemical Supplier No. 1755110 MG
A potent inhibitor of human PDGFRβ (IC50 = 80 nM)