Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(DmpSR')RuCl(P-(4-Fluorophenyl)3)], TCI America™
SDP

Catalog No. C3328100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3328100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3328100MG Supplier TCI America Supplier No. C3328100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1420299-84-8
Molecular Formula C42H37ClF3PRuS
Molecular Weight (g/mol) 798.31
MDL Number MFCD30725745
InChI Key PCBRQBCMBTZBQC-UHFFFAOYSA-L
Synonym [(DmpSR′C)RuCl(P-(4-Fluorophenyl)3)]
PubChem CID 134128902
IUPAC Name [2',4',6'-trimethyl-3-(2,4,6-trimethylphenyl)-[1,1'-biphenyl]-2-yl]sulfanide; chlororutheniumylium; tris(4-fluorophenyl)phosphane
SMILES Cl[Ru+].FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1.CC1=CC(C)=C(C(C)=C1)C1=CC=CC(=C1[S-])C1=C(C)C=C(C)C=C1C

Specifications

Quantity 100 mg
Formula Weight 798.31
Physical Form Crystalline Powder
Chemical Name or Material Chloro[(1,2,3,4,5,6-eta)-2,2′′,4,4′′,6,6′′-hexamethyl[1,1′
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.