Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dmpu-Hf Reagent

Catalog No. 8027945G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-279-45G 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 80-279-45G Supplier Sigma Aldrich Supplier No. 8027945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DMPU-HF, like TREAT-HF and Olah's Reagent, is a convenient way of utilizing Hydrogen Fluoride as a fluorination reagent. However, unlike TREAT-HF and Olah's Reagent, DMPU-HF is more tolerant of transition metals due to the reduced basicity of DMPU, compared to Triethylamine and Pyridine. Hammond and coworkers display its compatibility in transition-metal catalysis in gold-catalyzed mono- and difluoroination of alkynes.Designer HF-Based Fluorination Reagent: Highly Regioselective Synthesis of Fluoroalkenes and gem-Difluoromethylene Compounds from Alkynes

Specifications

CAS 1391826-58-6
Melting Point 117.8°C
Molecular Formula C10H7N3O2
Linear Formula C10H7N3O2
MDL Number MFCD16652405
Quantity 10 mg
Molecular Weight (g/mol) 201.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.