Learn More
Aviva Systems Biology Corp Dmrt1 Antibody - C-terminal region (ARP37217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dmrt1. It was validated on Western Blot. Dmrt1,doublesex and mab-3 related transcription factor 1,Dmrt1,doublesex- and mab-3-related transcription factor 1,doublesex- and mab-3-related transcription factor 1The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: -ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-598