Learn More
Aviva Systems Biology Corp DMRT2 Antibody - N-terminal region (ARP40046_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40046_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DMRT2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DMRT2. Alternate names: ApplicationIHC, WB. Species reactivity: Human, Rabbit
Catalog No. 50-014-53170