Learn More
Aviva Systems Biology Corp DMRT3 Antibody - middle region (ARP33210_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33210_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMRT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DMRT3. Alternate names: DMRTA3, MGC142144ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63070