Learn More
Aviva Systems Biology Corp DMRTA1 Antibody - N-terminal region (ARP37557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMRTA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse DMRTA1. Alternate names: Dmrt4ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-53079