Learn More
Aviva Systems Biology Corp DMRTA2 Antibody - C-terminal region (ARP33214_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33214_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMRTA2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DMRTA2. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54920