Learn More
Aviva Systems Biology Corp DMRTB1 Antibody - middle region (ARP36093_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36093_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DMRTB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DMRTB1. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-56010