Learn More
Bioworld DMS, 1 g
Supplier:  Bioworld 218200021
Encompass_Preferred
Dimethyl suberimidate•2HClA longer spacer imidoester cross-linkerCategory: amine-reactiveReactive Groups: imidoesterReaticive Toward: amino groupsWater Solluble: YesMembrane Permeable: YesCleavability: Non-cleavableMW: 273.20
Catalog No. 50-199-278