Learn More
Bioworld DMSO, 100 mL
Catalog No. NC0533696
Encompass_Preferred
DMSO, Dimethyl SulfoxideTechnical Specifications:Grade: Molecular biologyForeign activity: DNase and RNase, none detectedAppearance: LiquidBoiling Point : 189 °C(lit.)Density : 1.10 g/mL(lit.)Melting Point: 16-19 °C(lit.)HS Code:2930.90.70
Catalog No. NC0533696 Supplier Bioworld Supplier No. 404700061
DMSO, Dimethyl SulfoxideTechnical Specifications:Grade: Molecular biologyForeign activity: DNase and RNase, none detectedAppearance: LiquidBoiling Point : 189 °C(lit.)Density : 1.10 g/mL(lit.)Melting Point: 16-19 °C(lit.)HS Code:2930.90.70