Learn More
Gold Biotechnology Inc DMSO ACS Grade 100 mL
SDP
Catalog No. 5000562112
Encompass_Preferred
Dimethylsulfoxide (DMSO) is a very polar organic compound that is useful in organic chemistry for solubilizing organic and inorganic compounds DMSO is also used as a cryoprotectant for cell media preventing the formation of ice crystals which would otherwise damage cells DMSO can be used in PCR reactions to inhibit the self-complementation of DNA or PCR primers and increase amplification especially for DNA sequences which have GC rich sequences
Catalog No. 50-005-62112 Supplier Gold Biotechnology Inc Supplier No. D360100
Dimethylsulfoxide (DMSO) is a very polar organic compound that is useful in organic chemistry for solubilizing organic and inorganic compounds DMSO is also used as a cryoprotectant for cell media preventing the formation of ice crystals which would otherwise damage cells DMSO can be used in PCR reactions to inhibit the self-complementation of DNA or PCR primers and increase amplification especially for DNA sequences which have GC rich sequences