Learn More
Gold Biotechnology Inc DMSO, ACS Grade 10 mL
SDP
Catalog No. NC2943333
Encompass_Preferred
Dimethylsulfoxide (DMSO) is a very polar organic compound that is useful in organic chemistry for solubilizing organic and inorganic compounds. DMSO is also used as a cryoprotectant for cell media, preventing the formation of ice crystals, which would otherwise damage cells. DMSO can be used in PCR reactions to inhibit the self-complementation of DNA or PCR primers and increase amplification, especially for DNA sequences, which have GC rich sequences.
Catalog No. NC2943333 Supplier Gold Biotechnology Inc Supplier No. D36010
Dimethylsulfoxide (DMSO) is a very polar organic compound that is useful in organic chemistry for solubilizing organic and inorganic compounds. DMSO is also used as a cryoprotectant for cell media, preventing the formation of ice crystals, which would otherwise damage cells. DMSO can be used in PCR reactions to inhibit the self-complementation of DNA or PCR primers and increase amplification, especially for DNA sequences, which have GC rich sequences.