Learn More
Cayman Chemical DMSO asy Reagent 1mL
SDP
Catalog No. 502896589
Encompass_Preferred
A DPP-9 inhibitor (IC50 0.0034 UM) selective for DDP-9 over DPP-8 DPP-4 DPP-2 FAP and prolyl endopeptidase (IC50s 0.6 >10 >5 10 and >10 UM respectively) induces LDH release from THP-1 and HL-60 leukemia cells at 10 UM
Catalog No. 50-289-6589 Supplier Cayman Chemical Supplier No. 7000011 ML
A DPP-9 inhibitor (IC50 0.0034 UM) selective for DDP-9 over DPP-8 DPP-4 DPP-2 FAP and prolyl endopeptidase (IC50s 0.6 >10 >5 10 and >10 UM respectively) induces LDH release from THP-1 and HL-60 leukemia cells at 10 UM