Learn More
Bioworld DMSO Bio-Max, Molecular Biology Grade, 67-68-5, 100ml
Supplier:  Bioworld 404700052
Encompass_Preferred
DMSO (Dimethyl sulfoxide) is a polar solvent used in many chemical reactions, solubilization of peptides, in polymerase chain reactions (PCR) and as a cryoprotectant agent for the preservation of cells. Biomax DMSO is used in cell freezing media to protect cells from ice crystal formation causing mechanical injury to the cells. DMSO is often used in the combinations with BSA or fetal bovine serum (FBS) in various media. Technical Specifications: Tissue Culture Grade, DMSO, Sterile filtered, Cell culture tested. For use in cell freezing. Appearance: Liquid, Boiling Point : 189 C(lit.), Density : 1.10 g/mL(lit.), Melting Point: 16-19 C(lit.), HS Code: 2930.90.70, Molecular Weight : 78.13, Molecular Formula : C2H6OS, Purity : >99.5%, Solubility : Soluble in water, ethanol, ether, chloroform, benzene, and acetone. Appearance Form : Liquid. Appearance Color : Colorless to faint yellow. Melting Point : 16 - 19C, Boiling Point : 189C, Density : 1.10 g/mL, 100ml
Catalog No. 50-255-891