Learn More
FUJIFILM Biosciences DMSOFRE proteFREE ANIMAL 10ML
Catalog No. 502892159
Encompass_Preferred
Eliminate the risk of DMSO toxicity and maintain potency of human mesenchymal stem/ stromal cells (MSCs), PBMCs (T cells), and hematopoietic stem cells (HSCs) throughout cryopreservationComparable post-thaw cell viability to solutions containing DMSONontoxic when injected in animal models (refer to toxicological application note)Enables cell preservation for short-term at -80°C* and long-term storage in liquid nitrogen to -196°CComplete, ready-to-use medium* Human MSC and PBMC (T cell) data available for short-term storage. Human HSC data is not available.By purchasing this product you are agree that you will not use this product as excipient
Catalog No. 50-289-2159 Supplier FUJIFILM Biosciences Supplier No. 9114010ML
Eliminate the risk of DMSO toxicity and maintain potency of human mesenchymal stem/ stromal cells (MSCs), PBMCs (T cells), and hematopoietic stem cells (HSCs) throughout cryopreservationComparable post-thaw cell viability to solutions containing DMSONontoxic when injected in animal models (refer to toxicological application note)Enables cell preservation for short-term at -80°C* and long-term storage in liquid nitrogen to -196°CComplete, ready-to-use medium* Human MSC and PBMC (T cell) data available for short-term storage. Human HSC data is not available.By purchasing this product you are agree that you will not use this product as excipient