Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dmt-2'-f-dc(ac) amidite | 159414-99-0 | 99.5% | C41H49FN5O8P | 1 G
SDP

Catalog No. 5000358249
Encompass_Preferred

Dmt-2'-f-dc(ac) amidite is a phosphoramidite utilized in the preparation of cyclic purine dinucleotides. It is provided as a white to off-white solid, ensuring a high purity of 99.5%. This product is intended for research purposes only.

  • Used in the preparation of cyclic purine dinucleotides.
  • High purity of 99.5%.
  • Appearance: White to off-white solid.
  • Recommended storage for solid form: -20°C, sealed, away from moisture and light.
  • Recommended storage for solutions: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture and light.

Catalog No. 50-003-58249 Supplier Medchemexpress LLC Supplier No. HY454911G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dmt-2'-f-dc(ac) amidite is a phosphoramidite utilized in the preparation of cyclic purine dinucleotides. It is provided as a white to off-white solid, ensuring a high purity of 99.5%. This product is intended for research purposes only.

  • Used in the preparation of cyclic purine dinucleotides.
  • High purity of 99.5%.
  • Appearance: White to off-white solid.
  • Recommended storage for solid form: -20°C, sealed, away from moisture and light.
  • Recommended storage for solutions: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.