Learn More
Medchemexpress LLC Dmt-2'-f-dc(ac) amidite | 159414-99-0 | MFCD12912404 | 99.5% | C41H49FN5O8P | 1 ML
SDP
Catalog No. 5000359091
Encompass_Preferred
Dmt-2'-f-dc(ac) amidite, also known as 2'-F-Ac-dC Phosphoramidite, is a phosphoramidite utilized in the synthesis of cyclic purine dinucleotides.
  • Used in oligonucleotide synthesis
  • Applied in cancer, blood, and cardio-cerebrovascular disease research
  • Functions as a nucleoside antimetabolite/analog
  • Relevant for DNA/RNA synthesis studies
Catalog No. 50-003-59091 Supplier Medchemexpress LLC Supplier No. HY4549110MM/1ML
Dmt-2'-f-dc(ac) amidite, also known as 2'-F-Ac-dC Phosphoramidite, is a phosphoramidite utilized in the synthesis of cyclic purine dinucleotides.
  • Used in oligonucleotide synthesis
  • Applied in cancer, blood, and cardio-cerebrovascular disease research
  • Functions as a nucleoside antimetabolite/analog
  • Relevant for DNA/RNA synthesis studies