Learn More
Medchemexpress LLC DMT-2 -F-iBu-G 10mM 1mL
SDP
Catalog No. 5000206111
Encompass_Preferred
DMT-2 -F-iBu-G (5 -DMT-2 -F-ibu-dG) is a modified oligonucleotide 2 -deoxy-2 -fluoro-modified oligonucleotides shows high nuclease resistant and retained exceptional binding affinity to the RNA targets[1]
Catalog No. 50-002-06111 Supplier Medchemexpress LLC Supplier No. HYW11530910MM/1ML
DMT-2 -F-iBu-G (5 -DMT-2 -F-ibu-dG) is a modified oligonucleotide 2 -deoxy-2 -fluoro-modified oligonucleotides shows high nuclease resistant and retained exceptional binding affinity to the RNA targets[1]