Learn More
Medchemexpress LLC DMT-2'fluoro-da(bz) amidite | 136834-22-5 | 99.6% | 875.92 | 100 MG
SDP
Catalog No. 5000400649
Encompass_Preferred
DMT-2'fluoro-da(bz) amidite is a key intermediate used for synthesizing antisense oligonucleotides that exhibit high nuclease resistance, strong RNA binding affinity, and maintained base-pair specificity.
  • Key intermediate for synthesizing antisense oligonucleotides.
  • Confers high nuclease resistance to antisense oligonucleotides.
  • Provides high RNA binding affinity.
  • Maintains base-pair specificity.
  • 2'-fluoro modification enhances the stability of hybrid duplex.
  • 2'-fluoro modification makes the duplex present an A-form conformation.
Catalog No. 50-004-00649 Supplier Medchemexpress LLC Supplier No. HY21997100MG
DMT-2'fluoro-da(bz) amidite is a key intermediate used for synthesizing antisense oligonucleotides that exhibit high nuclease resistance, strong RNA binding affinity, and maintained base-pair specificity.
  • Key intermediate for synthesizing antisense oligonucleotides.
  • Confers high nuclease resistance to antisense oligonucleotides.
  • Provides high RNA binding affinity.
  • Maintains base-pair specificity.
  • 2'-fluoro modification enhances the stability of hybrid duplex.
  • 2'-fluoro modification makes the duplex present an A-form conformation.