Learn More
Medchemexpress LLC DMT-2'Fluoro-DG(IB) Amidite | 144089-97-4 | 99.5% | C44H53FN7O8P | 100 MG
SDP
Catalog No. 5000358350
Encompass_Preferred
DMT-2'Fluoro-DG(IB) Amidite is a modified nucleoside phosphoramidite with a fluorine on the 2' position, designed for oligonucleotide synthesis. It can also be used in the preparation of 4'-modified 2'-deoxy-2'-fluorouridine.
  • Can be employed for oligonucleotide synthesis.
  • Can be used to prepare 4'-modified 2'-deoxy-2'-fluorouridine.
  • Modified nucleoside phosphoramidite with a fluorine on the 2' position.
  • For research use only.
Catalog No. 50-003-58350 Supplier Medchemexpress LLC Supplier No. HY45492100MG
DMT-2'Fluoro-DG(IB) Amidite is a modified nucleoside phosphoramidite with a fluorine on the 2' position, designed for oligonucleotide synthesis. It can also be used in the preparation of 4'-modified 2'-deoxy-2'-fluorouridine.
  • Can be employed for oligonucleotide synthesis.
  • Can be used to prepare 4'-modified 2'-deoxy-2'-fluorouridine.
  • Modified nucleoside phosphoramidite with a fluorine on the 2' position.
  • For research use only.