Learn More
Sigma Aldrich Fine Chemicals Biosciences DMT-2'O-Methyl-rC(ac) Phosphoramidite configured for PerkinElmer, configured for Polygen | 199593-09-4 | 0.5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C2131800.5G
Encompass
DMT-2'O-Methyl-rC(ac) Phosphoramidite configured for PerkinElmer, configured for Polygen | Purity: >=99% (31P-NMR); >=99.0% (reversed phase HPLC); conforms (1H-NMR); conforms (LC/MS) | Mol Wt: 801.86 | 199593-09-4 | 0.5G
Catalog No. 50-188-4354