Learn More
Medchemexpress LLC DMT-2'O-Methyl-rC(tac) Phosphoramidite | 179486-26-1 | 95.0% | 950.07 | 250 MG
SDP
Catalog No. 5000402917
Encompass_Preferred
DMT-2'O-Methyl-rC(tac) phosphoramidite is a phosphoramidite primarily used in the synthesis of oligonucleotides. This product is suitable for laboratory research in areas such as DNA/RNA synthesis, cell cycle, and DNA damage studies.
  • Can be used in the synthesis of oligonucleotides
  • Suitable for DNA/RNA synthesis research
  • Appears as a solid, white to light yellow in color
  • Recommended storage at 4°C under nitrogen
Catalog No. 50-004-02917 Supplier Medchemexpress LLC Supplier No. 250MGHY21713
DMT-2'O-Methyl-rC(tac) phosphoramidite is a phosphoramidite primarily used in the synthesis of oligonucleotides. This product is suitable for laboratory research in areas such as DNA/RNA synthesis, cell cycle, and DNA damage studies.
  • Can be used in the synthesis of oligonucleotides
  • Suitable for DNA/RNA synthesis research
  • Appears as a solid, white to light yellow in color
  • Recommended storage at 4°C under nitrogen