Learn More
Medchemexpress LLC DMT-2 O-Methyl-rU Ph 10mM 1mL
SDP
Catalog No. 5000205476
Encompass_Preferred
DMT-2 O-Methyl-rU Phosphoramidite (2 -O-Me-U Phosphoramidite) is a 2 -O-Me derivative and can be used for oligonucleotide synthesis[1][2]
Catalog No. 50-002-05476 Supplier Medchemexpress LLC Supplier No. HYW10139110MM/1ML
DMT-2 O-Methyl-rU Phosphoramidite (2 -O-Me-U Phosphoramidite) is a 2 -O-Me derivative and can be used for oligonucleotide synthesis[1][2]